Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrin alpha 1 Rabbit anti-Human, Clone: SPTA1/2939R, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$304.50

Specifications

Antigen Spectrin alpha 1
Clone SPTA1/2939R
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, ELISA 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122004
SDP
View Documents
Novus Biologicals
NBP30726520UG
20 μg
Each of 1 for $304.50
Only null left
Add to Cart
 
Description

Description

Spectrin alpha 1 Monoclonal specifically detects Spectrin alpha 1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry-Paraffin.
Specifications

Specifications

Spectrin alpha 1
0.2 mg/ml
Western Blot, ELISA, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cell Biology, Cellular Markers
10 mM PBS with 0.05% BSA
6708
IgG
Protein A or G purified
SPTA1/2939R
Western Blot 1-2 ug/ml, ELISA 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Monoclonal
Purified
RUO
Human
alpha-I spectrin, EL2, erythrocyte, HS3, spectrin, alpha, erythrocytic 1 (elliptocytosis 2)
Recombinant fragment of human Spectrin alpha 1 (around aa356-475) (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.