Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrin alpha chain, non-erythrocytic 1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980239
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0239 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0239 Supplier Bioss Supplier No. BS0381R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPTAN1 (spectrin alpha chain, non-erythrocytic 1) is a part of the spectrin family of filamentous cytoskeletal proteins. They function as essential scaffold proteins that stabilize the plasma membrane and organize intracellular organelles. Spectrins are composed of alpha and beta dimers that associate to form tetramers linked in a head-to-head arrangement. SPTAN1 has been implicated in other cellular functions including DNA repair and cell cycle regulation. Mutations in this gene are the cause of early infantile epileptic encephalopathy-5. Alternate splicing results in multiple transcript variants.

Specifications

Antigen Spectrin alpha chain, non-erythrocytic 1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SPTAN1
Gene Accession No. Q13813
Gene Alias (ALPHA)II-SPECTRIN; 2610027H02Rik; A2a; a--fodrin; alpha II spectrin; alpha2 spectrin; alpha-fodrin; Alpha-II spectrin; alpha-spectrin 2; alpha-spectrin 2, brain; BOS_12120; brain alpha-spectrin; cardiomyocyte alpha-spectrin; EIEE5; FLJ44613; Fodrin alpha chain; im:7157190; inhibitory protein factor; IPF; LOC100049693; NEAS; noerythroid alpha-spectrin 2; nonerythroid alpha-spectrin 2; sequence with a GC signal; Spectrin alpha; spectrin alpha 2; spectrin alpha chain, brain; Spectrin alpha chain, non-erythrocytic 1; spectrin alpha non-erythrocytic 1; spectrin alpha, non-erythrocytic 1; spectrin, alpha, non-erythrocytic 1; spectrin, alpha, non-erythrocytic 1 (alpha-fodrin); spectrin, non-erythroid alpha chain; Spectrin, non-erythroid alpha subunit; Spna2; Spna-2; Spta2; Sptan1; Unknown (protein for IMAGE:7150239); Unknown (protein for MGC:112229); wu:fa20e05; wu:fb33g08; wu:fk32a10; zgc:112229; zgc:112229 protein
Gene Symbols SPTAN1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Spectrin alpha chain, non-erythrocytic 1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6709
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.