Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Spectrin beta 3 Antibody (1287A) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP280958
Description
Spectrin beta 3 Monoclonal specifically detects Spectrin beta 3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).Specifications
Spectrin beta 3 | |
Monoclonal | |
1 mg/mL | |
Western Blot : 2 μg/mL, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence : 5 - 20 μg/mL, Immunohistochemistry-Paraffin : 1:1000, Flow (Intracellular) : 1 μg/mL | |
Beta-III spectrin, glutamate transporter EAAT4-associated protein 41, GTRAP41, KIAA0302, SCA5, spectrin beta chain, brain 2, spectrin, beta, non-erythrocytic 2, Spectrin, non-erythroid beta chain 2, spinocerebellar ataxia 5 | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry | |
1287A | |
Unconjugated | |
PBS | |
SPTBN2 | |
Synthetic peptide containing the C-terminal region of human Spectrin beta 3 (UniProt O15020) | |
0.1 mg | |
Neuroscience | |
6712 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction