Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Spectrin beta 3 Antibody (SPTBN2/1582), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254488PECY55
Description
Spectrin beta 3 Monoclonal specifically detects Spectrin beta 3 in Human samples. It is validated for Flow Cytometry.Specifications
Spectrin beta 3 | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
SPTBN2 | |
Recombinant fragment (around aa356-475) of human Spectrin beta 3 protein (exact sequence is proprietary) (Uniprot: O15020) | |
0.1 mL | |
Primary | |
Human | |
IgG2a κ |
Flow Cytometry | |
SPTBN2/1582 | |
Flow Cytometry | |
Beta-III spectrin, glutamate transporter EAAT4-associated protein 41, GTRAP41, KIAA0302, SCA5, spectrin beta chain, brain 2, spectrin, beta, non-erythrocytic 2, Spectrin, non-erythroid beta chain 2, spinocerebellar ataxia 5 | |
Mouse | |
Protein A or G purified | |
Neuroscience | |
6712.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction