Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Spectrin beta-3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558724
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558724 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558724 Supplier Invitrogen™ Supplier No. PA558724
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PPPSTQAPSV NGVCTDGEPS QPLLGQQRLE HSSFPEGPGP GSGDEANGPR GERQTRTRGP APSAMPQSRS TESAH Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 79%.

Spectrins are principle components of a cell's membrane-cytoskeleton and are composed of two alpha and two beta spectrin subunits. The protein encoded by this gene (SPTBN2), is called spectrin beta non-erythrocytic 2 or beta-III spectrin. It is related to, but distinct from, the beta-II spectrin gene which is also known as spectrin beta non-erythrocytic 1 (SPTBN1). SPTBN2 regulates the glutamate signaling pathway by stabilizing the glutamate transporter EAAT4 at the surface of the plasma membrane. Mutations in this gene cause a form of spinocerebellar ataxia, SCA5, that is characterized by neurodegeneration, progressive locomotor incoordination, dysarthria, and uncoordinated eye movements.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Spectrin beta-3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Sptbn2
Gene Accession No. O15020
Gene Alias beta SpIII sigma 1; beta-III spectrin; beta-spectrin 3; glutamate transporter EAAT4-associated protein 41; GTRAP41; KIAA0302; mKIAA0302; SCA5; SCAR14; Spectrin Beta; spectrin beta 3; spectrin beta chain, brain 2; spectrin beta chain, non-erythrocytic 2; spectrin beta III sigma 2; spectrin beta, non-erythrocytic 2; spectrin, beta, non-erythrocytic 2; spectrin, non-erythroid beta chain 2; spectrin-like protein GTRAP41; spinocerebellar ataxia 5 protein; Spnb3; SPNB-3; SPTBN2
Gene Symbols Sptbn2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Spectrin beta-3. Recombinant protein control fragment (Product #RP-103917).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6712
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.