Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Spectrin beta-4 Monoclonal Antibody (N393/2), APC
GREENER_CHOICE

Catalog No. PIMA545721
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545721 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545721 Supplier Invitrogen™ Supplier No. MA545721
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

A 1:100 dilution of MA5-45721 was sufficient for detection of Beta 4 Spectrin in 20 μg of mouse brain lysate by ECL immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately >200kDa. Does not cross react with other Beta-spectrins This antibody was formerly sold as clone S393-2.

Spectrin is the major constituent of the cytoskeletal network underlying the erthrocyte plasma membrane and determines cell shape, arrangement of transmembrane proteins and organization of organelles. It is expressed at very low levels in many tissues, with the strongest expression in the cerebellum, spinal cord, stomach, pituitary gland, liver, pancreas, salivary gland, kidney, bladder and heart. Spectrin beta 4 is a non-erythrocyte member, and is expressed in the brain and pancreatic islets and localisted to the nuclear matrix, cytoplasmic vesicles and PML nuclear bodies. Beta4 spectrins are also essential for membrane stability and the molecular organization of the nodes of Ranvier.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Spectrin beta-4
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N393/2
Concentration 1 mg/mL
Conjugate APC
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene SPTBN4
Gene Accession No. Q9H254
Gene Alias 1700022P15Rik; 5830426A08Rik; beta-IV spectrin; beta-spectrin 4; dyn; EMBL BAB83243.1; KIAA1642; lnd; lumbosacral neuroaxonal dystrophy; neuroaxonal dystrophy; nmf261; nmf379; quivering; qv; ROSA62; SpbIV; spectrin beta 4; spectrin beta chain, brain 3; spectrin beta chain, non-erythrocytic 4; spectrin beta, non-erythrocytic 4; spectrin, beta, non-erythrocytic 4; spectrin, non-erythroid beta chain 3; SPNB4; SPTBN3; SPTBN4
Gene Symbols SPTBN4
Host Species Mouse
Immunogen Fusion protein amino acids 1621-1832 (C-terminal repeats 14 to 15) of human Beta4-spectrin.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 308458, 57731, 80297
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.