Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Spectrin beta-4 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5121581
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121581 20 μL
PIPA5121580 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121581 Supplier Invitrogen™ Supplier No. PA5121581
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Spectrin is the major constituent of the cytoskeletal network underlying the erthrocyte plasma membrane and determines cell shape, arrangement of transmembrane proteins and organization of organelles. It is expressed at very low levels in many tissues, with the strongest expression in the cerebellum, spinal cord, stomach, pituitary gland, liver, pancreas, salivary gland, kidney, bladder and heart. Spectrin beta 4 is a non-erythrocyte member, and is expressed in the brain and pancreatic islets and localisted to the nuclear matrix, cytoplasmic vesicles and PML nuclear bodies. Beta4 spectrins are also essential for membrane stability and the molecular organization of the nodes of Ranvier.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Spectrin beta-4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene SPTBN4
Gene Accession No. Q9H254
Gene Alias 1700022P15Rik; 5830426A08Rik; beta-IV spectrin; beta-spectrin 4; dyn; EMBL BAB83243.1; KIAA1642; lnd; lumbosacral neuroaxonal dystrophy; neuroaxonal dystrophy; nmf261; nmf379; quivering; qv; ROSA62; SpbIV; spectrin beta 4; spectrin beta chain, brain 3; spectrin beta chain, non-erythrocytic 4; spectrin beta, non-erythrocytic 4; spectrin, beta, non-erythrocytic 4; spectrin, non-erythroid beta chain 3; SPNB4; SPTBN3; SPTBN4
Gene Symbols SPTBN4
Host Species Rabbit
Immunogen Lys64-Ser285 (Accession Q9H254), with N-terminal His Tag.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57731
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.