Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Spectrin beta-5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140081
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140081 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140081 Supplier Invitrogen™ Supplier No. PA5140081
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: EKVASIALLD LTGARCERLR GRHGRKHTFS LRLTSGAEIL FAAPSEEQAE SWWRALGSTA AQSLSPKLKA KPVSSL Highest antigen sequence indentity to the following orthologs: Mouse - 32%, Rat - 32%.

SPTBN5 (Spectrin Beta, Non-Erythrocytic 5) is a Protein Coding gene. Diseases associated with SPTBN5 include Epileptic Encephalopathy, Early Infantile, 5 and Spinocerebellar Ataxia 5. Among its related pathways are RET signaling and Signaling by GPCR. Gene Ontology (GO) annotations related to this gene include actin binding and kinesin binding. An important paralog of this gene is DST. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Spectrin beta-5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPTBN5
Gene Accession No. Q9NRC6
Gene Alias beta V spectrin; beta-V spectrin; BSPECV; HU HUSPECV; HUBSPECV; HUSPECV; spectrin beta chain, brain 4; spectrin beta chain, non-erythrocytic 5; spectrin beta, non-erythrocytic 5; spectrin, beta, non-erythrocytic 5; spectrin, non-erythroid beta chain 4; SPTBN5
Gene Symbols SPTBN5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPTBN5. Recombinant protein control fragment (Product #RP-108919).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51332
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.