Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Spectrin beta-5 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550141

Catalog No. PA550141


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total SPTBN5 protein.

SPTBN5 (Spectrin Beta, Non-Erythrocytic 5) is a Protein Coding gene. Diseases associated with SPTBN5 include Epileptic Encephalopathy, Early Infantile, 5 and Spinocerebellar Ataxia 5. Among its related pathways are RET signaling and Signaling by GPCR. Gene Ontology (GO) annotations related to this gene include actin binding and kinesin binding. An important paralog of this gene is DST. [GeneCards].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Spectrin beta-5
Polyclonal
Unconjugated
SPTBN5
beta V spectrin; beta-V spectrin; BSPECV; HU HUSPECV; HUBSPECV; HUSPECV; spectrin beta chain, brain 4; spectrin beta chain, non-erythrocytic 5; spectrin beta, non-erythrocytic 5; spectrin, beta, non-erythrocytic 5; spectrin, non-erythroid beta chain 4; SPTBN5
Rabbit
Antigen affinity chromatography
RUO
51332
-20°C
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9NRC6
SPTBN5
Synthesized peptide derived from internal of human SPTBN5.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.