Learn More
Spectroline™ Camera Adapters for GeneLine Image Documentation Systems
SDP
Catalog No. 11992342 Shop All Spectronics Corporation Products
Change view
Click to view available options
Adap For Cc-80,ccd,geneline,ea
Adap For Cc-80,dig,geneline,ea
Adap Forcl-150,ccd,geneline,ea
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
11-992-342 Adap For Cc-80,dig,geneline,ea
11-992-345 Adap For Cc-80,ccd,geneline,ea
11-992-346 Adap Forcl-150,ccd,geneline,ea
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-992-342 Supplier Spectroline™ Supplier No. GL1300
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Includes
Mounting brackets
  • AdaptGeneLine CCD or digital cameras for use with FisherBiotech™ UV viewing cabinets
  • GeneLine components are available separately for mix-and-match systems based on your individual needs