Learn More
Spectroline™ ImageAide Autoanalysis Software
SDP
Includes: Acquisition Board, CCD Camera, cables, and software
Products 2
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
11-992-349
SDP
View Documents
Spectroline™
GL2500
Software, Upgrade,imageaide,ea Software, Upgrade,imageaide,ea N/A N/A N/A
11-992-351
SDP
View Documents
Spectroline™
GL3200
Software,analysis,imageaide,ea Software,analysis,imageaide,ea N/A N/A N/A
Description
Description
  • For customizable analysis of digital images of all gel media, colonies, spots, and blots
  • Imports camera images and automatically finds lanes, bands, and edges, producing track profiles and results in less than ten seconds
  • Determines molecular weights, quantity, and Rf, then displays all in a single window; also allows data export directly into an Excel™ spreadsheet and Microsoft™ Word™ document
  • Autotrack finding feature detects distorted or misshapen tracks and bands
  • Supplied with the Spectroline™ GeneLine, ImageEZE, DigiCAM, or Epi-DigiCAM series; available also as separate analysis software package
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.