Learn More
Spectroline™ ImageAide Autoanalysis Software
SDP
Supplier:  Spectroline™ GL3200
 View more versions of this product
Catalog No. 11-992-351
Includes:
Acquisition Board, CCD Camera, cables, and software
Description
Description
  • For customizable analysis of digital images of all gel media, colonies, spots, and blots
  • Imports camera images and automatically finds lanes, bands, and edges, producing track profiles and results in less than ten seconds
  • Determines molecular weights, quantity, and Rf, then displays all in a single window; also allows data export directly into an Excel™ spreadsheet and Microsoft™ Word™ document
  • Autotrack finding feature detects distorted or misshapen tracks and bands
  • Supplied with the Spectroline™ GeneLine, ImageEZE, DigiCAM, or Epi-DigiCAM series; available also as separate analysis software package
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.