Learn More
Bolle Safety Spectrum Goggle
With its bi-material frame and adjustable strap, Spectrum offers superior protection and adaptability. Available in two sizes, Spectrum is a versatile product with different frame types and lens coatings to provide a solution for every company, regardless of their specific work environment. Its sporty design and optimized ergonomics also ensure greater acceptance for enhanced safety and productivity.
Supplier: Bolle Safety SPECTN14W
Description
- PC/TPR Co-injected frame for optimum comfort.
- Multiple coatings and sizes available.
- Soft gasket to fit most face structures.
- Optimal panoramic vision.
Specifications
Platinum | |
Safety Goggles | |
Polycarbonate | |
Polycarbonate | |
None | |
Not Autoclavable |
Clear | |
SPECTN14W | |
Reusable | |
Polycarbonate | |
Indirect Vented |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.