Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrum™ Spectra Mesh™ Woven, Nylon Filters
SDP

Catalog No. p-4707080
Click to view available options
Pore Size:
10 μm
20 μm
41 μm
53 μm
100 μm
Diameter (Metric):
47 mm
55 mm
90 mm
Thickness (Metric):
45 μm
55 μm
60 μm
78 μm

Consist of individual strands woven into a mesh screen

Combines high tensile strength with flexibility

  • Water Affinity: Hydrophillic
  • Strength: High
  • Adsorption: High
  • Best Resistance: Corrosives
  • pH Resistance: pH 3-10
  • Thermal Stability: up to 180°C
  • Sterilization: Irradiation

Specifications

Format Disc
Material Nylon
Particle Retention 100 μm
Pore Size 100 μm
For Use With (Application) Hazardous waste, drinking water purification, Acid rain measurement, suspended soils, air/asbestos, coal dust analysis, oceanography, trace element analysis
Thickness (Metric) 78 μm
Wettability Hydrophilic
Diameter (Metric) 90 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.