Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrum™ Spectra/Por™ Biotech Cellulose Ester (CE) Dialysis Membrane Tubing
SDP

Catalog No. p-2387504
Click to view available options
Diameter (Metric):
6.4 mm
10 mm
15 mm
20 mm
24 mm
MWCO:
0.1 to 0.5 kDa
0.5 kDa
1 kDa
100 kDa
25 kDa
5 kDa
50 kDa
This item is not returnable. View return policy
This item is not returnable. View return policy

Selected MWCOs from 100 to 100,000 Daltons

  • CE tubing membranes are ideal for desalting, purification, and molecular separation
  • Supplied in 10m (33 ft.) lengths in two rolls of 5m each, in 0.1% sodium azide solution

Order Info

Note:Do not tie—must be used with Spectra/Por nylon closures (08-671-52, -53, -55). Strong polar solvents such as acetone, MEK, or dioxane will irreparably damage membranes. Contact Spectrum Technical Service, 1-800-634-3300, for more information.

Specifications

Product Type Spectra/Por Dialysis Membrane Tubing
MWCO 0.5 kDa
Material Cellulose Ester
Diameter (Metric) 20 mm
Diameter (English) 0.78 in.
Length (English) 33 ft.
Length (Metric) 10 m
Formulation Supplied in 0.1% sodium azide solution
Temperature Range (English) 39.2°F to 98.6°F
Temperature Range (Metric) 4°C to 37°C
Quantity Each
For Use With (Application) Ideal for desalting, purification, and molecular separation
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.