Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrum™ Standard RC Dialysis Tubing
SDP

Catalog No. p-4394892
Click to view available options
Diameter (Metric):
6.4 mm
16 mm
20.4 mm
48 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

Multipurpose dialysis tubing is ideal for general dialysis for a variety of applications and volumes

  • Membrane grade
  • Membrane type: RC
  • Molecular weight cut-off: 12 to 14K

Specifications

Product Type Dialysis Membrane
MWCO 12 to 14 kDa
Material Regenerated Cellulose
Diameter (Metric) 48 mm
Diameter (English) 1.88 in.
Length (English) 50 ft.
Length (Metric) 15 m
Capacity (Metric) 18 mL
Quantity 50 Pack
For Use With (Application) Some tubing rolls do not contain glycerin and do not require soaking prior to use
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.