Learn More
Eisco™ Spectrum Tubes
SCI_ED
Catalog No. p-7122745
Click to view available options
Product Type:
Krypton Spectrum Tube
Spectrum Tube, Argon
Spectrum Tube, Carbon Dioxide
Spectrum Tube, Helium
Spectrum Tube, Hydrogen
Spectrum Tube, Iodine
Spectrum Tube, Neon
Spectrum Tube, Nitrogen
Spectrum Tube, Oxygen
Spectrum Tube, Sulphur
Spectrum Tube, Xenon
Includes
Spectrum Tube
For demonstrating spectral lines of pure substances.
  • Features large 10cm capillary in center for greater viewing area
  • For use with power supplies holding 26cm spectrum tubes
  • Designed for operation intervals of 30 seconds
  • Should be used for 30 seconds on/30 seconds off
For intermittent use only, not to be used as a lamp source.
Specifications
Product Type Spectrum Tube, Argon
For Use With (Application) Demonstrates the spectral lines of pure substances
Includes Spectrum Tube, large 10cm capillary
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.