Learn More
3M™ Speedglas™ 9100V Auto Darkening Filter 06-0000-10, Shades 5 and 8-13
The Speedglas 9100V filter is designed for the professional welder incorporating dark shade ranges 8-13, grinding and torch cutting modes, and advanced arc detection electronics for use with all arc welding processes.
Supplier:  3M™ 7010385085
Catalog No. 17-000-893
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
  • 1.80 in. x 3.70 in. ADF Viewing AreaSolar assist.
  • Features grinding and cutting modes.
  • Selectable Shades 5, 8-13.
  • Helps reduce eye strain.
  • The Speedglas 9100V filter designed for the professional welder incorporating dark shade ranges 8-13, grinding and torch cutting modes, and advanced arc detection electronics for use with all arc welding processes.
  • Viewing area 6.05 sq. in.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2800 hours
CR2032
9100 V
Auto-darkening Filter
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.