Learn More
3M™ Speedglas™ 9100XXi Auto Darkening Filter 06-0000-30i, Shades 5 and 8-13
Auto Darkening Filter 9100XXi offers the same large viewing area as our 3M™ Speedglas™ ADF 9100XX. But thanks to improved optics, the 9100XXi ADF allows you to more readily recognize colors as compared to our other models.
Supplier:  3M™ 7010342472
Catalog No. 17-000-891
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Grab and go with auto-on.
  • Two memory modes that allow you to set individual settings for dark shade, light-to-dark switching sensitivity, and dark-to-light delay (Requires silver front panel 9100XXi).
  • External grinding mode activation (Requires silver front panel 9100XXi).
  • 9100XXi ADF fits with all 3M™ Speedglas™ Welding Helmet Series 9100. However, it requires the silver front panel 9100XXi to utilize the grinding and memory outside control functions.
  • This silver front panel 9100XXi only fits 3M™ Speedglas™ Welding.
  • Helmets 9100/9100-Air/9100 QR.
  • An improved user interface that is easy to understand.
  • 3M™ Speedglas™ Auto Darkening Filter 9100XXi offers a 2.8 in. x 4.2 in. viewing area (approx).
  • Selectable Shades 5, 8-13 and Shade 3 light state.
  • Delay function to set the dark-to-light opening time.
  • Sensitivity of the arc detection that can be adjusted.
  • Comfort mode for tack welding and two-step recovery are not available with this model.
  • When used with a compatible 3M™ Speedglas™ Welding Helmet Series 9100, meets ANSI Z87.1-2010 high impact.
  • Silver front panel 9100XXi sold separately.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ADF: up to 1800 Hours
ADF: CR2032
9100 XXi
Auto-darkening Filter
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.