Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Speedglas™ FA III SAR Welding System

Catalog No. 17000892 Shop All 3M Products
Change view
Click to view available options
Certifications/Compliance:
NIOSH, ANSI Z87.1-2010
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
17-000-892 NIOSH, ANSI Z87.1-2010
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-000-892 Supplier 3M™ Supplier No. 7010342473
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Supplied air respiratory (SAR) system includes V-100 Vortex air-cooling valve, complete Speedglas™ Welding Helmet 9100-Air, and 9100XXi ADF.

  • Welding respiratory protection system featuring the V-100 cool-air valve
  • Lightweight belt-mounted valve allows welders to lower temperature of air entering headpiece by up to 50°F (28°C); designed for hot, strenuous jobs
  • With improved optics, Auto Darkening Filter 9100XXi allows colors to be recognized more readily as compared to other models
  • Grab and go with auto-on feature
  • 9100XXi silver front panel houses external controls, enabling fast and easy access to the grinding and memory modes
  • Two memory modes that allow you to set individual settings for dark shade, light-to-dark switching sensitivity, and dark-to-light delay
  • External grinding mode activation
  • Improved user interface is easy to understand
  • Auto Darkening Filter 9100XXi offers a 2.8 in. x 4.2 in. viewing area (approx)
  • Selectable Shades 5, 8-13 and Shade 3 light state
  • Delay function to set the dark-to-light opening time
  • Sensitivity of the arc detection can be adjusted
  • Comfort mode for tack welding and two-step recovery are not available with this model
  • Side windows - Shade 5 side filters that help increase peripheral vision.
  • Ergonomic, adjustable head suspension for increased comfort, helmet stability and weight distribution
  • NIOSH-approved system; meets ANSI Z87.1-2010 high impact
  • Storage bag, welding cap, and starter kit included
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance NIOSH, ANSI Z87.1-2010
Color Black
Product Type Supplied Air Respirator Welding System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.