Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories SpeedySwab™ COVID-19/FLU AB Rapid Test

Catalog No. 21381509
Change view
Click to view available options
Covid-19; Flu AB; Nasal Swab
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
21-381-509 Covid-19; Flu AB; Nasal Swab
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-381-509 Supplier Germaine™ Laboratories Supplier No. LFA046225N

missing translation for 'validMainframeMsg'

Lateral flow immunochromatographic assay intended for in-vitro rapid, simultaneous qualitative detection and differentiation of influenza A and B nucleoprotein antigens and SARS CoV-2 nucleocapsid antigen.

  • Lateral flow immunochromatographic assay intended for in-vitro rapid, simultaneous qualitative detection and differentiation of influenza A and B nucleoprotein antigens and SARS CoV-2 nucleocapsid antigen
  • Sample directly from anterior nasal swab specimens of individuals with signs and symptoms of respiratory infection consistent with COVID-19 within the first five days of symptom onset when tested at least twice over three days with at least 48 hours between tests
  • This test is FDA/EUA authorized for use at the Point of Care, i.e., in patient care settings operating under a CLIA Certificate of Waiver, Certificate of Compliance, or Certificate of Accreditation.

Specifications

CE Marker No
Clia Complexity Waived (EUA)
Detectable Analytes Influenza A, Influenza B, COVID-19/2019-nCOV
Detection Method Simultaneous Qualitative Detection
Format Kit
Sample Type Single Nasal Swab Sample
Sensitivity COVID Clinical Sensitivity: 92.6%;, FLU-A Clinical Sensitivity: 82.9%, FLU-B Clinical Sensitivity: 90.0%, Refer to the Package Insert for additional performance claims
Type Rapid Test
Disposable Single-use
For Use With (Application) Covid and Flu AB Rapid Test
Form Ready-to-Use (RTU)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.