Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPEG Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553875
Description
Immunogen sequence: VYKSVIANKL GKAACYAHLY VTDVVPGPPD GAPQVVAVTG RMVTLTWNPP RSLDMAIDPD SLTYTVQHQV Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 99%.

Specifications
SPEG | |
Polyclonal | |
Unconjugated | |
SPEG | |
aortic preferentially expressed gene 1; aortic preferentially expressed protein 1; APEG1; APEG-1; BPEG; CNM5; KIAA1297; nuclear protein, marker for differentiated aortic smooth muscle and down-regulated with vascular injury; SPEG; SPEG complex locus; SPEGalpha; SPEGbeta; striated muscle preferentially expressed protein kinase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10290 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15772 | |
SPEG | |
Recombinant protein corresponding to Human SPEG. Recombinant protein control fragment (Product #RP-92276). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction