Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPEM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144156
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144156 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144156 Supplier Invitrogen™ Supplier No. PA5144156
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPEM1 Polyclonal Antibody for IHC (P), ELISA

SPEM1 is required for proper cytoplasm removal during spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPEM1
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene SPEM1
Gene Accession No. Q8N4L4
Gene Alias C17orf83; SPEM1; spermatid maturation 1; spermatid maturation protein 1
Gene Symbols SPEM1
Host Species Rabbit
Immunogen Recombinant Human Spermatid maturation protein 1 protein (50-309AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374768
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.