Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPEN Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10058798AF700
Description
SPEN Polyclonal antibody specifically detects SPEN in Human samples. It is validated for Western BlotSpecifications
SPEN | |
Polyclonal | |
Western Blot | |
HIAA0929, KIAA0929nuclear receptor transcription cofactor, MINTMsx2 interacting nuclear target (MINT) homolog, RBM15C, SHARPmsx2-interacting protein, SMART/HDAC1 associated repressor protein, SMART/HDAC1-associated repressor protein, SPEN homolog, spen homolog, transcriptional regulator (Drosophila) | |
The immunogen recognized by this antibody maps to a region between residue 1350 and 1400 of human SMART/HDAC1 associated repressor protein (spen homolog, transcriptional regulator) using the numbering given in entry NP_055816.2 (GeneID 23013). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
23013 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction