Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPEN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565605
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565605 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565605 Supplier Invitrogen™ Supplier No. PA565605
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GEAVENQEVQS KKPIPSKPQL KQLQVLDDQG PEREDVRKNY CSLRDETPER KSGQEKSHSV NTEEKIGIDI DHTQSYRKQM EQSRRKQQME MEIAKSEKFG SPKKDVDEYE RRSLVHEVGK PPQDVTDDSP PSK Highest antigen sequence identity to the following orthologs - mouse 78%, rat 82%.

May serve as a nuclear matrix platform that organizes and integrates transcriptional responses. In osteoblasts, supports transcription activation: synergizes with RUNX2 to enhance FGFR2-mediated activation of the osteocalcin FGF-responsive element (OCFRE). Has also been shown to be an essential corepressor protein, which probably regulates different key pathways such as the Notch pathway. Negative regulator of the Notch pathway via its interaction with RBPSUH, which prevents the association between NOTCH1 and RBPSUH, and therefore suppresses the transactivation activity of Notch signaling. Blocks the differentiation of precursor B-cells into marginal zone B-cells. Probably represses transcription via the recruitment of large complexes containing histone deacetylase proteins. May bind both to DNA and RNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPEN
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPEN
Gene Accession No. Q96T58
Gene Alias HIAA0929; KIAA0929; MINT; Msx2 interacting nuclear target (MINT) homolog; msx2-interacting protein; nuclear receptor transcription cofactor; RBM15C; SHARP; SMART/HDAC1-associated repressor protein; SPEN; spen family transcriptional repressor; SPEN homolog; spen homolog, transcriptional regulator
Gene Symbols SPEN
Host Species Rabbit
Immunogen Recombinant Human SPEN. Recombinant protein control fragment (Product #RP-106268).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23013
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.