Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Sperian™ Freedom Disposable Respirators

Catalog No. 19821396
Click to view available options
Filter Type:
Multi-contaminant (organic vapors, acid gas, ammonia, methylamine, formaldehyde)
Organic Vapor (OV)
Organic vapor w/R95
Includes:
Attached ov cartridge, r95 prefilter, retainer
Filter Pad, Organic Vapor Cartridge, Retainer, Reusable Bag
Filter Pad, Retainer, Reusable Bag
Reusable Bag
Size:
Medium
Large
Suspension Type:
Yolk and cradle

Ideal for occasional users

  • Available in multiple models for a variety of applications
  • Three sizes ensure a secure, comfortable fit
  • Choose from contaminant-specific models or multi-contaminant, multi-use respirators (with cartridge and filter pre-assembled), designed to meet NIOSH standards for multiple contaminant exposure
  • Dispose when cartridges are expended, replacement parts and decontamination concerns are eliminated

Order Info

See chart below for a complete selection of respirators with pre-attached cartridges and filters. Probed models are also available; contact your Fisher Scientific™ Customer Service Representative for details. (Do not exceed maximum use concentration established by regulatory standards.)

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance NIOSH 42CFR84
Color Code Blue
Disposable Single-use
Filter Type Organic vapor w/R95
Material Thermoplastic Elastomer
Product Type Half Mask Respirator
Size Large
For Use With (Application) Fire protection, medical, pharmaceutical, transportation
Includes Attached ov cartridge, r95 prefilter, retainer
Material (Faceshield) Thermoplastic Elastomer
Nosepiece Type Forward angled
Quantity 1 Ea.
Suspension Type Yolk and cradle
Weight (English) 0.75 lb.
Weight (Metric) 0.34 kg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.