Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ SAF-T-FIT™ Plus N95 Particulate Respirators

Catalog No. 19034908 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
With Exhalation Valve:
No
Yes
Color:
Gray
Red
White
Product Type:
General Purpose Particulate Respirator
Particulate Respirator with Nuisance Level Acid Gas Relief
Size:
Medium
Medium/Large
Small
X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. With Exhalation Valve Color Product Type Size
19-034-908 Yes Red Particulate Respirator with Nuisance Level Acid Gas Relief Small
19-005-165 No White General Purpose Particulate Respirator Medium
19-013-822 Yes White General Purpose Particulate Respirator X-Large
19-013-820 Yes White General Purpose Particulate Respirator Small
19-013-821 Yes White General Purpose Particulate Respirator Medium
19-034-909 Yes Gray Particulate Respirator with Nuisance Level Acid Gas Relief Medium/Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-034-908 Supplier Honeywell™ Supplier No. 14110399

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

View Alternate Product

This item is not returnable. View return policy

For use with particulate aerosols free of oil

  • Low breathing resistance
  • Incorporates humidity-resistant filter media
  • Durable suspension straps
  • Packaged to maintain integrity of product in work environment
  • Adjustable boomerang nose seal for secure fit
  • Color coded nose clips
  • Latex-free

TRUSTED_SUSTAINABILITY

Specifications

Aerosol Type N - Not Resistant to Oil
Certifications (NIOSH) NIOSH 42 CFR 84, N95, NIOSH approval number: TC-84A-3789
Color Red
Design Type Cup/Molded
Disposable Single-use
Filter Type N95
Product Type Particulate Respirator with Nuisance Level Acid Gas Relief
Size Small
With Exhalation Valve Yes
Filter Efficiency 95%
For Use With (Application) Welding, soldering, brazing, torch cutting, metal pouring, grinding, sanding
Latex Free Yes
Model 14110399
Quantity 10/Pk.
Strap Attachment Type Two-Strap Design
FDA Cleared Surgical Mask No
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.