Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sperm Protein sp56 Monoclonal Antibody (5F12), QED Bioscience™
SDP

Catalog No. 502278127
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8127 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-8127 Supplier Leinco Technologies Inc Supplier No. 55102100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sperm Protein sp56 Monoclonal antibody specifically detects Sperm Protein sp56 in Hamster, Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen Sperm Protein sp56
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5F12
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Zp3r
Gene Accession No. Q60736
Gene Alias AC122773.1; AV264399; SP56; sperm fertilization protein 56; zona pellucida 3 receptor; zona pellucida sperm-binding protein 3 receptor; Zp3r
Host Species Mouse
Immunogen Mixture of native and formaldehyde-zinc fixed mouse sperm purified through a Percoll gradient
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22789
Target Species Hamster, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.