Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Spermine synthase Antibody, Novus Biologicals™

Chicken Polyclonal Antibody
Supplier: Novus Biologicals NB10075148
Description
Spermine synthase Polyclonal specifically detects Spermine synthase in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Spermine synthase | |
Polyclonal | |
Western Blot 1:100-1:2000 | |
P52788 | |
SMS | |
The immunogen for this antibody was a sequence from 206a.a. to 335a.a. from Spermine synthase(a 368a.a. long protein) | |
0.05 mg | |
Core ESC Like Genes, Stem Cell Markers | |
6611 | |
Human, Mouse, Rat |
Western Blot | |
Unconjugated | |
PBS with No Preservative | |
EC 2.5.1.22, MRSRSpS, Snyder-Robinson X-linked mental retardation syndrome, Spermidine aminopropyltransferase, spermine synthase, SPMSYSRS | |
Chicken | |
Affinity Purified | |
RUO | |
Primary | |
Spermine synthase | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction