Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPERT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558752
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558752 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558752 Supplier Invitrogen™ Supplier No. PA558752
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NKALREENRM LSKENKILQV FWEEHKASLG REESRAPSPL LHKDSASLEV VKKDHVALQV PRGKEDSTLQ LLREENRALQ QLLEQKQAY Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 71%.

SPERT (spermatid associated protein), also known as CBY2 (chibby homolog 2) or NURIT, is a novel leucine-zipper protein belonging to the chibby family of proteins. Uniquely Expressed in the spermatid flower-like structure, SPERT interacts with Nek1, a member of the NIMA-family kinase family that is associated centrosomal stability and ciliogenesis. It contains a leucine-zipper motif and two coiled-coil regions, and is transcribed through the elongation stage of the spermatids. SPERT is absent from mature spermatozoa and is thought to be involved in transporting proteins that are to be discarded via the residual bodies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPERT
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPERT
Gene Accession No. Q8NA61
Gene Alias 1700086N05Rik; AI596460; CBY2; chibby family member 2; chibby homolog 2; novel leucine zipper testicular protein; Nurit; Protein chibby homolog 2; protein nurit; spermatid associated; spermatid flower-like structure protein; spermatid-associated protein; Spert; testicular tissue protein Li 182; testis-specific leucine zipper protein nurit
Gene Symbols SPERT
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPERT. Recombinant protein control fragment (Product #RP-97989).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 220082
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.