Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPERT Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559043
Description
Immunogen sequence: ASQHSYPLNR FSSVPLDPME RPMSQADLEL DYNPPRVQLS DEMFVFQDGR WVNENCRLQS PYFSPSASFH HKLHHK Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 93%.
SPERT (spermatid associated protein), also known as CBY2 (chibby homolog 2) or NURIT, is a novel leucine-zipper protein belonging to the chibby family of proteins. Uniquely Expressed in the spermatid flower-like structure, SPERT interacts with Nek1, a member of the NIMA-family kinase family that is associated centrosomal stability and ciliogenesis. It contains a leucine-zipper motif and two coiled-coil regions, and is transcribed through the elongation stage of the spermatids. SPERT is absent from mature spermatozoa and is thought to be involved in transporting proteins that are to be discarded via the residual bodies.
Specifications
SPERT | |
Polyclonal | |
Unconjugated | |
SPERT | |
1700086N05Rik; AI596460; CBY2; chibby family member 2; chibby homolog 2; novel leucine zipper testicular protein; Nurit; Protein chibby homolog 2; protein nurit; spermatid associated; spermatid flower-like structure protein; spermatid-associated protein; Spert; testicular tissue protein Li 182; testis-specific leucine zipper protein nurit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
220082 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NA61 | |
SPERT | |
Recombinant protein corresponding to Human SPERT. Recombinant protein control fragment (Product #RP-97990). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction