Learn More
SPF30 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302837AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, X. tropicalis, Bovine and Orangutan
This gene is a paralog of SMN1 gene, which encodes the survival motor neuron protein, mutations in which are cause of autosomal recessive proximal spinal muscular atrophy. The protein encoded by this gene is a nuclear protein that has been identified as a constituent of the spliceosome complex. This gene is differentially expressed, with abundant levels in skeletal muscle, and may share similar cellular function as the SMN1 gene.Specifications
SPF30 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O75940, Q8BGT7 | |
Smndc1 | |
Between 188 and 238 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
SMNDC1 | |
SMNR, SPF30, 30 kDa splicing factor SMNrp, SMN-related protein, splicing factor 30, survival of motor neuron-related, survival motor neuron domain-containing protein 1, survival of motor neuron-related-splicing factor 30 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10285, 76479 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.