Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPFH2 Antibody (SR1636), Novus Biologicals™
SDP

Catalog No. p-200005390 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259513 100 μg
NB258775 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB259513 Supplier Novus Biologicals Supplier No. NBP322439100UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SPFH2 Monoclonal antibody specifically detects SPFH2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence

Specifications

Antigen SPFH2
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone SR1636
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias C8orf2SPFH domain-containing protein 2, Endoplasmic reticulum lipid raft-associated protein 2, ER lipid raft associated 2, Erlin-2, MGC87072, NET32, SPFH domain family, member 2, SPFH2chromosome 8 open reading frame 2, Stomatin-prohibitin-flotillin-HflC/K domain-containing protein 2
Host Species Rabbit
Immunogen A synthesized peptide derived from human SPFH2 (Uniprot #: O94905)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Alzheimers Research, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11160
Target Species Human, Mouse
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.