Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPFH2 Rabbit anti-Human, Mouse, Rat, Clone: 9T3M5, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
Antigen | SPFH2 |
---|---|
Clone | 9T3M5 |
Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
SPFH2 Monoclonal antibody specifically detects SPFH2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
SPFH2 | |
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
C8orf2SPFH domain-containing protein 2, Endoplasmic reticulum lipid raft-associated protein 2, ER lipid raft associated 2, Erlin-2, MGC87072, NET32, SPFH domain family, member 2, SPFH2chromosome 8 open reading frame 2, Stomatin-prohibitin-flotillin-HflC/K domain-containing protein 2 | |
A synthetic peptide corresponding to a sequence within amino acids 50-150 of human SPFH2 (O94905). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
9T3M5 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Alzheimers Research, Neurodegeneration, Neuroscience, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
11160 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title