Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPG20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106297
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106297 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106297 Supplier Invitrogen™ Supplier No. PA5106297
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Spartin, SPG20.

SPG20 is a protein containing a MIT (Microtubule Interacting and Trafficking molecule) domain, and is implicated in regulating endosomal trafficking and mitochondria function. The protein localizes to mitochondria and partially co-localizes with microtubules. Stimulation with epidermal growth factor (EGF) results in protein translocation to the plasma membrane, and the protein functions in the degradation and intracellular trafficking of EGF receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPG20
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SPART
Gene Accession No. Q8N0X7, Q8R1X6
Gene Alias AI840044; C79168; KIAA0610; mKIAA0610; Spart; Spartin; Spartin-like protein; spastic paraplegia 20 (Troyer syndrome); spastic paraplegia 20 (Troyer syndrome) homolog; Spastic paraplegia 20 protein; spastic paraplegia 20, spartin (Troyer syndrome) homolog; spastic paraplegia 20, spartin (Troyer syndrome) homolog (human); SPG20; TAHCCP1; Trans-activated by hepatitis C virus core protein 1
Gene Symbols SPART, Spg20
Host Species Rabbit
Immunogen A synthesized peptide derived from human SPART(Accession Q8N0X7), corresponding to amino acid residues H401-K451.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 229285, 23111, 295053
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.