Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPG20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110421
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110421 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110421 Supplier Invitrogen™ Supplier No. PA5110421
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEQEPQNGEP AEIKIIREAY KKAFLFVNKG LNTDELGQKE EAKNYYKQGI GHLLRGISIS SKESEHTGPG WESARQMQQK MKETLQNVRT RLEILEKGLA TSLQNDLQEV PKLYPEFPPK DMCEKLPEPQ SFSSAPQHAE VNGNTSTPSA GAVAAPASLS LPSQSCPAEA PPAYTPQAAE GHYTVSYGTD SGEFSSVGEE FYRNHSQPPP LETLGLDADE LILIPNGVQI FFVNPAGEVS APSYPGYLRI VRFLDNSLDT VLNRPPGFLQ VCDWLYPLVP DRSPVLKCTA GAYMFPDTML QAAGCFVGVV LSSELPEDDR ELFEDLLRQM SDLRLQANWN RAEEENEFQI.

SPG20 is a protein containing a MIT (Microtubule Interacting and Trafficking molecule) domain, and is implicated in regulating endosomal trafficking and mitochondria function. The protein localizes to mitochondria and partially co-localizes with microtubules. Stimulation with epidermal growth factor (EGF) results in protein translocation to the plasma membrane, and the protein functions in the degradation and intracellular trafficking of EGF receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPG20
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SPART
Gene Accession No. Q8N0X7, Q8R1X6
Gene Alias AI840044; C79168; KIAA0610; mKIAA0610; Spart; Spartin; Spartin-like protein; spastic paraplegia 20 (Troyer syndrome); spastic paraplegia 20 (Troyer syndrome) homolog; Spastic paraplegia 20 protein; spastic paraplegia 20, spartin (Troyer syndrome) homolog; spastic paraplegia 20, spartin (Troyer syndrome) homolog (human); SPG20; TAHCCP1; Trans-activated by hepatitis C virus core protein 1
Gene Symbols SPART, Spg20
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-350 of human SPG20 (NP_055902.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 229285, 23111, 295053
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.