Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPG20 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544282
Description
Peptide sequence: ASWVSWGLVK GAEITGKAIQ KGASKLRERI QPEEKPVEVS PAVTKGLYIA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.
SPG20 is a protein containing a MIT (Microtubule Interacting and Trafficking molecule) domain, and is implicated in regulating endosomal trafficking and mitochondria function. The protein localizes to mitochondria and partially co-localizes with microtubules. Stimulation with epidermal growth factor (EGF) results in protein translocation to the plasma membrane, and the protein functions in the degradation and intracellular trafficking of EGF receptor.
Specifications
SPG20 | |
Polyclonal | |
Unconjugated | |
SPART | |
AI840044; C79168; KIAA0610; mKIAA0610; Spart; Spartin; Spartin-like protein; spastic paraplegia 20 (Troyer syndrome); spastic paraplegia 20 (Troyer syndrome) homolog; Spastic paraplegia 20 protein; spastic paraplegia 20, spartin (Troyer syndrome) homolog; spastic paraplegia 20, spartin (Troyer syndrome) homolog (human); SPG20; TAHCCP1; Trans-activated by hepatitis C virus core protein 1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
23111 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N0X7 | |
SPART | |
Synthetic peptide directed towards the middle region of human SPG20. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction