Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPG20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558638
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558638 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558638 Supplier Invitrogen™ Supplier No. PA558638
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AGAYMFPDTM LQAAGCFVGV VLSSELPEDD RELFEDLLRQ MSDLRLQANW NRAEEENEFQ IPGRTRPSSD QLKEASGTDV KQLDQGNKDV RHKGKRGKRA KDTSSE Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 74%.

SPG20 is a protein containing a MIT (Microtubule Interacting and Trafficking molecule) domain, and is implicated in regulating endosomal trafficking and mitochondria function. The protein localizes to mitochondria and partially co-localizes with microtubules. Stimulation with epidermal growth factor (EGF) results in protein translocation to the plasma membrane, and the protein functions in the degradation and intracellular trafficking of EGF receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPG20
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPART
Gene Accession No. Q8N0X7
Gene Alias AI840044; C79168; KIAA0610; mKIAA0610; Spart; Spartin; Spartin-like protein; spastic paraplegia 20 (Troyer syndrome); spastic paraplegia 20 (Troyer syndrome) homolog; Spastic paraplegia 20 protein; spastic paraplegia 20, spartin (Troyer syndrome) homolog; spastic paraplegia 20, spartin (Troyer syndrome) homolog (human); SPG20; TAHCCP1; Trans-activated by hepatitis C virus core protein 1
Gene Symbols SPART
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPG20. Recombinant protein control fragment (Product #RP-97948).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23111
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.