Learn More
Invitrogen™ SPG21 Monoclonal Antibody (2B11)

Description
Peptide Sequence: PHKIRDIPVT IMDVFDQSAL STEAKEEMYK LYPNARRAHL KTGGNFPYLC RSAEVNLYVQ IHLLQFHGTK YAAIDPSMVS AEELEVQKGS LGISQE.
Specifications
Specifications
| Antigen | SPG21 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 2B11 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | SPG21 |
| Gene Accession No. | Q9NZD8 |
| Gene Alias | acid cluster protein 33; ACP33; BM-019; C78576; D9Wsu18e; GL010; Maspardin; MAST; spastic paraplegia 21 (autosomal recessive, Mast syndrome); Spastic paraplegia 21 autosomal recessive Mast syndrome protein; Spastic paraplegia 21 autosomal recessive Mast syndrome protein homolog; spastic paraplegia 21 homolog (human); Spastic paraplegia 21 protein; SPG21 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.