Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPG3A Antibody (1F6B12) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | SPG3A |
---|---|
Clone | 1F6B12 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61886
![]() |
Novus Biologicals
NBP261886 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
SPG3A Monoclonal specifically detects SPG3A in Human samples. It is validated for Western Blot, ELISA.Specifications
SPG3A | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
51062 | |
Purified recombinant fragment of human ATL1 (AA: 1-100) expressed in E. Coli. | |
Primary | |
63.5 kDa |
1F6B12 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
AD-FSP, atlastin GTPase 1, atlastin1, atlastin-1, Brain-specific GTP-binding protein, EC 3.6.5.-, FSP1, GBP3, GBP-3, GTP-binding protein 3, Guanine nucleotide-binding protein 3, guanylate-binding protein 3, Spastic paraplegia 3 protein A, spastic paraplegia 3A (autosomal dominant), SPG3, SPG3AhGBP3 | |
ATL1 | |
IgG1 | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title