Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPG48 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981647
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1647 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1647 Supplier Bioss Supplier No. BS11760R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The function of this protein has not been specifically defined.

Specifications

Antigen SPG48
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene AP5Z1
Gene Accession No. O43299
Gene Alias adapter-related protein complex 5 zeta subunit; adaptor related protein complex 5 zeta 1 subunit; adaptor related protein complex 5, zeta 1 subunit; Adaptor-related protein complex 5 zeta subunit; adaptor-related protein complex 5, zeta 1 subunit; AP-5 complex subunit zeta-1; AP5Z1; KIAA0415; protein KIAA0415; SPG48; zeta; Zeta5
Gene Symbols AP5Z1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SPG48 .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9907
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.