Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPG7 Antibody (OTI1G4), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274323JF646
Description
SPG7 Monoclonal specifically detects SPG7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SPG7 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SPG7 | |
Full length recombinant protein of human SPG7 (NP_003110) produced in E.coli. | |
0.1 mL | |
6687.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI1G4 | |
Western Blot, Immunohistochemistry | |
CARMGC126331, cell matrix adhesion regulator, CMARMGC126332, EC 3.4.24, EC 3.4.24.-, FLJ37308, paraplegin, PGNcell adhesion regulator, spastic paraplegia 7 (pure and complicated autosomal recessive), Spastic paraplegia 7 protein, SPG5C | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction