Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPG7 Monoclonal Antibody (OTI1C12)
GREENER_CHOICE

Catalog No. PIMA525907
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525907 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525907 Supplier Invitrogen™ Supplier No. MA525907
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SPG7 Monoclonal Antibody for Western Blot, ICC/IF, Flow

This gene encodes a mitochondrial metalloprotease protein that is a member of the AAA family. Members of this protein family share an ATPase domain and have roles in diverse cellular processes including membrane trafficking, intracellular motility, organelle biogenesis, protein folding, and proteolysis. Mutations in this gene cause autosomal recessive spastic paraplegia 7. Two transcript variants encoding distinct isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPG7
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI1C12
Concentration 0.86 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SPG7
Gene Accession No. Q3ULF4, Q9UQ90
Gene Alias AI452278; AU015315; CAR; cell adhesion regulator; cell matrix adhesion regulator; CMAR; paraplegin; paraplegin, isoform 1; PGN; spastic paraplegia 7 (pure and complicated autosomal recessive); spastic paraplegia 7 homolog (human); spastic paraplegia 7 protein; SPG5C; SPG7; SPG7 matrix AAA peptidase subunit, paraplegin; SPG7, paraplegin matrix AAA peptidase subunit
Gene Symbols SPG7
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 300-573 of SPG7 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 234847, 6687
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.