Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPG7 Monoclonal Antibody (OTI1D2), TrueMAB™, OriGene
SDP

Catalog No. 502540335
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0335 30 μL
50-167-5117 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0335 Supplier OriGene Supplier No. TA504423S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SPG7 Monoclonal antibody specifically detects SPG7 in Human samples. It is validated for Western Blot

Specifications

Antigen SPG7
Applications Western Blot
Classification Monoclonal
Clone OTI1D2
Concentration 0.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene HLCS
Gene Accession No. P50747
Gene Alias AI452278; AU015315; CAR; cell adhesion regulator; cell matrix adhesion regulator; CMAR; paraplegin; paraplegin, isoform 1; PGN; spastic paraplegia 7 (pure and complicated autosomal recessive); spastic paraplegia 7 homolog (human); spastic paraplegia 7 protein; SPG5C; SPG7; SPG7 matrix AAA peptidase subunit, paraplegin; SPG7, paraplegin matrix AAA peptidase subunit
Host Species Mouse
Immunogen Full length human recombinant protein of human HLCS produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3141
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.