Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPG8 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP282036AF405
This item is not returnable.
View return policy
Description
SPG8 Polyclonal antibody specifically detects SPG8 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
SPG8 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA | |
Alexa Fluor 405 | |
KIAA0196, MGC111053, spastic paraplegia 8 (autosomal dominant), SPG8, strumpellin, WASH complex subunit strumpellin | |
SPG8 antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human SPG8. The immunogen is located within the last 50 amino acids of SPG8. Amino Acid Squence: DYVRYTKLPRRVAEAHVPN | |
0.1 mL | |
Neuroscience | |
9897 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction