Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Spherical Pinch Clamp
GREENER_CHOICESDP

Spherical Pinch Clamp features 304 Stainless Steel construction with a screw type lock nut.

$19.90 - $210.24

Specifications

Type Pinch Clamp
Material Stainless Steel
Includes Screw Down Locking Nut
For Use With (Application) Organic Synthesis
Color Silver
View More Specs

Products 7
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
31-501-930
GREENER_CHOICESDP
View Documents
Synthware™
CC0012
12/5, #5
Each for $19.90
Only null left
Add to Cart
 
31-501-931
GREENER_CHOICESDP
View Documents
Synthware™
CC0018
18/8, #9
Each for $27.72
Only null left
Add to Cart
 
31-501-932
GREENER_CHOICESDP
View Documents
Synthware™
CC2815
28/15, #15
Each for $29.58
Only null left
Add to Cart
 
31-501-933
GREENER_CHOICESDP
View Documents
Synthware™
CC3525
35/20, #25
Each for $38.04
Only null left
Add to Cart
 
31-501-934
GREENER_CHOICESDP
View Documents
Synthware™
CC0050
50/30, #30
Each for $115.68
Only null left
Add to Cart
 
31-501-935
GREENER_CHOICESDP
View Documents
Synthware™
CC0065
65/40, #40
Each for $115.68
Only null left
Add to Cart
 
31-501-936
GREENER_CHOICESDP
View Documents
Synthware™
CC0075
75/50, #50
Each for $210.24
Only null left
Add to Cart
 
Description

Description

  • With a screw type locking nut.
Specifications

Specifications

Pinch Clamp
Screw Down Locking Nut
Silver
Stainless Steel
Organic Synthesis
Spring Loaded
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.