Learn More
Description
- Dispense into working aliquots if needed; repeated freeze-thaws will destroy product integrity.
- Source: Murine Engelbreth-Holm-Swarm (EHS) tumor and bovine extensor tendon.
Specifications
Specifications
| Content And Storage | Product is stable for a minimum of 3 months from date of shipment. Store at -80° C; keep frozen. Dispense into working aliquots if needed; repeated freeze-thaws will destroy product integrity. |
| Product Line | R&D Systems |
| Quantity | 6 mL |
| Stability | Product is stable for a minimum of 3 months from date of shipment. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
