Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sphingosine 1 phosphate phosphatase 2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB151766 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151766 25 μg
NB151765 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB151766 Supplier Novus Biologicals Supplier No. NBP31190625UG
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sphingosine 1 phosphate phosphatase 2 Polyclonal antibody specifically detects Sphingosine 1 phosphate phosphatase 2 in Human samples. It is validated for Western Blot

Specifications

Antigen Sphingosine 1 phosphate phosphatase 2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:400
Formulation Whole antiserum
Gene Alias EC 3.1.3, EC 3.1.3.-, FLJ39004, hSPP2, sphingosine 1-phosphate phosphohydrolase 2, Sphingosine-1-phosphatase 2, sphingosine-1-phosphate phosphatase 2, Spp2, SPP2sphingosine-1-phosphate phosphotase 2, SPPase2
Host Species Rabbit
Immunogen Synthetic peptide derived from the Sphingosine 1 phosphate phosphatase 2 protein. (Uniprot: Q8IWX5)
Purification Method Unpurified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130367
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.