Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sphingosine Kinase 1/SPHK1 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP122974C
Description
Sphingosine Kinase 1/SPHK1 Polyclonal specifically detects Sphingosine Kinase 1/SPHK1 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Sphingosine Kinase 1/SPHK1 | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
Q2M3C7 | |
SPHK1 | |
sphingosine kinase 1 | |
Affinity Purified | |
RUO | |
Primary | |
Human, Rat | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
EC 2.7.1.91, sphingosine kinase 1, SPHKSK 1, SPK, SPK 1 | |
Rabbit | |
42.5 kDa | |
0.1 mL | |
Lipid and Metabolism | |
8877 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction