Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sphingosine Kinase 1/SPHK1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP323415
Description
Sphingosine Kinase 1/SPHK1 Polyclonal antibody specifically detects Sphingosine Kinase 1/SPHK1 in Human samples. It is validated for Western BlotSpecifications
Sphingosine Kinase 1/SPHK1 | |
Polyclonal | |
Western Blot 1:1000 | |
EC 2.7.1.91, sphingosine kinase 1, SPHKSK 1, SPK, SPK 1 | |
Sphingosine Kinase 1/SPHK1 synthetic peptide (coupled to carrier protein) corresponding to an unphosphorylated sequence of amino acids surrounding serine 225 in the central region of human Sphingosine Kinase 1/SPHK1. This sequence has 4 amino acid differences from mouse and 5 from rat Sphingosine Kinase 1/SPHK1, and is not homologous to sequences in Sphingosine Kinase 2/SPHK2. [UniProt# Q9NYA1] | |
0.1 mL | |
Lipid and Metabolism | |
8877 | |
Store at -20°C. | |
IgG |
Western Blot | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction